Tisk…

5-Methylpicolinic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: 5-Methylpyridine-2-carboxylic acid

OR301172-1G OR301172-5G
APOSOR301172-1GEA 0 CZK
APOSOR301172-1G APOSOR301172-5G
5-Methylpicolinic acid
5-Methylpicolinic acid
Vzorec: C₇H₇NO₂
MW: 137.1360 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01704358
CAS číslo: 4434-13-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR