Tisk…

2-Amino-5-ethyl-pyridine

Dodavatel: Apollo Scientific

OR301037-1G
APOSOR301037-1GEA 6910 CZK
APOSOR301037-1G
2-Amino-5-ethyl-pyridine
2-Amino-5-ethyl-pyridine
Vzorec: C₇H₁₀N₂
Storage Temperature: Ambient
MDL Number: MFCD09757479
CAS číslo: 19842-07-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR