Tisk…

Ethyl 3-(4-benzylpiperidin-1-yl)propionate

Dodavatel: Apollo Scientific

OR300893-1G OR300893-5G
APOSOR300893-1GEA 0 CZK
APOSOR300893-1G APOSOR300893-5G
Ethyl 3-(4-benzylpiperidin-1-yl)propionate
Ethyl 3-(4-benzylpiperidin-1-yl)propionate
Vzorec: C₁₇H₂₅NO₂
Storage Temperature: Ambient
MDL Number: MFCD13633484
CAS číslo: 1099610-66-8

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR