Tisk…

Methylaminophenylacetonitrile hydrochloride

Dodavatel: Apollo Scientific

OR300787-10G OR300787-1G OR300787-5G
APOSOR300787-10GEA 0 CZK
APOSOR300787-10G APOSOR300787-1G APOSOR300787-5G
Methylaminophenylacetonitrile hydrochloride
Methylaminophenylacetonitrile hydrochloride
Vzorec: C₉H₁₁ClN₂
Storage Temperature: Ambient
MDL Number: MFCD11052442
CAS číslo: 5537-52-0

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR