Tisk…

1-Bromo-3-(2,2-dibromovinyl)-5-isopropylbenzene

Dodavatel: Apollo Scientific
Varování

OR300225-250MG
APOSOR300225-250MGEA 8570 CZK
APOSOR300225-250MG
1-Bromo-3-(2,2-dibromovinyl)-5-isopropylbenzene
1-Bromo-3-(2,2-dibromovinyl)-5-isopropylbenzene
Vzorec: C₁₁H₁₁Br₃
MW: 382,917 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31561256
CAS číslo: 2227272-84-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR