Tisk…

1-(Isoquinolin-3-yl)ethanone

Dodavatel: Apollo Scientific

OR300098-250MG OR300098-1G
APOSOR300098-250MGEA 2690 CZK
APOSOR300098-250MG APOSOR300098-1G
1-(Isoquinolin-3-yl)ethanone
1-(Isoquinolin-3-yl)ethanone
Vzorec: C₁₁H₉NO
Storage Temperature: Ambient
MDL Number: MFCD06658291
CAS číslo: 91544-03-5

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR