Tisk…

2-Chloroisonicotinic acid hydrazide

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Chloro-4-pyridinecarbohydrazide

OR29934-1G OR29934-250MG OR29934-5G
APOSOR29934-1GEA 1910 CZK
APOSOR29934-1G APOSOR29934-250MG APOSOR29934-5G
2-Chloroisonicotinic acid hydrazide
2-Chloroisonicotinic acid hydrazide
Vzorec: C₆H₆ClN₃O
MW: 171.59 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00221428
CAS číslo: 58481-04-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR