Tisk…

4-Methoxybenzyl isocyanate 98%

Dodavatel: Apollo Scientific
Nebezpečí

OR2873-1G
APOSOR2873-1GEA 0 CZK
APOSOR2873-1G
4-Methoxybenzyl isocyanate 98%
4-Methoxybenzyl isocyanate

  • May form small amount of sediment.

Vzorec: C₉H₉NO₂
Bod varu: 60…66 °C/0,6mmHg
Hustota: 1,143 g/cm³ (25 °C)
Bod vzplanutí: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00673064
CAS číslo: 56651-60-6
UN: 2206
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR