Tisk…

5-Chloro-2-nitroaniline

Dodavatel: Apollo Scientific
Nebezpečí

OR28701-100G OR28701-10G OR28701-25G
APOSOR28701-100GEA 1660 CZK
APOSOR28701-100G APOSOR28701-10G APOSOR28701-25G
5-Chloro-2-nitroaniline
5-Chloro-2-nitroaniline
Vzorec: C₆H₅ClN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00007776
CAS číslo: 1635-61-6
EINECS: 216-661-5
UN: 2237
ADR: 6.1,III

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR