Tisk…

Propagyl amine hydrochloride

Dodavatel: Apollo Scientific

Synonyms: Propargylamine hydrochloride, Prop-2-yn-1-amine hydrochloride

OR27447-25G
APOSOR27447-25GEA 6250 CZK
APOSOR27447-25G
Propagyl amine hydrochloride
Propagyl amine hydrochloride
Vzorec: C₃H₆ClN
MW: 91.54
Storage Temperature: Ambient
MDL Number: MFCD00012907
CAS číslo: 15430-52-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR