Tisk…

4-[(2,2-Dimethylpropanoyl)amino]nicotinic acid

Dodavatel: Apollo Scientific

OR25946-1G OR25946-250MG OR25946-500MG OR25946-2.5G OR25946-5G
APOSOR25946-1GEA 7840 CZK
APOSOR25946-1G APOSOR25946-250MG APOSOR25946-500MG APOSOR25946-2.5G APOSOR25946-5G
4-[(2,2-Dimethylpropanoyl)amino]nicotinic acid
4-[(2,2-Dimethylpropanoyl)amino]nicotinic acid
Vzorec: C₁₁H₁₄N₂O₃
Storage Temperature: Ambient
MDL Number: MFCD03407344
CAS číslo: 125867-31-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR