Tisk…

Ethyl 3-amino-4-cyano-5-(methylthio)thiophene-2-carboxylate

Dodavatel: Apollo Scientific

OR25286-1G
APOSOR25286-1GEA 4370 CZK
APOSOR25286-1G
Ethyl 3-amino-4-cyano-5-(methylthio)thiophene-2-carboxylate
Ethyl 3-amino-4-cyano-5-(methylthio)thiophene-2-carboxylate
Vzorec: C₉H₁₀N₂O₂S₂
Storage Temperature: Ambient
MDL Number: MFCD00085050
CAS číslo: 116170-90-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR