Tisk…

Ethyl 5-methyl-3-(methylthio)-4,4-dioxo-4,5-dihydro-4lambda~6~-benzo[c]thieno[3,4-e][1,2]thiazine-1-

Dodavatel: Apollo Scientific

OR25277-1G OR25277-5G
APOSOR25277-1GEA 2480 CZK
APOSOR25277-1G APOSOR25277-5G
Ethyl 5-methyl-3-(methylthio)-4,4-dioxo-4,5-dihydro-4lambda~6~-benzo[c]thieno[3,4-e][1,2]thiazine-1-
Ethyl 5-methyl-3-(methylthio)-4,4-dioxo-4,5-dihydro-4lambda~6~-benzo[c]thieno[3,4-e][1,2]thiazine-1-
Vzorec: C₁₅H₁₅NO₄S₃
Storage Temperature: Ambient
MDL Number: MFCD00085073
CAS číslo: 175276-42-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR