Tisk…

Trichloroacrylic acid

Dodavatel: Apollo Scientific
Nebezpečí

OR23621-1G OR23621-250MG
APOSOR23621-1GEA 15940 CZK
APOSOR23621-1G APOSOR23621-250MG
Trichloroacrylic acid
Trichloroacrylic acid
Vzorec: C₃HCl₃O₂
Bod tání: 72…74 °C
Storage Temperature: Ambient
MDL Number: MFCD00015455
CAS číslo: 2257-35-4
EINECS: 218-856-0
UN: 3261
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR