Tisk…

2,5-Dibromoterephthalic acid

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 2,5-Dibromoterephtalic acid

OR22280-1G OR22280-5G OR22280-10G
APOSOR22280-1GEA 622 CZK
APOSOR22280-1G APOSOR22280-5G APOSOR22280-10G
2,5-Dibromoterephthalic acid
2,5-Dibromoterephthalic acid
Vzorec: C₈H₄Br₂O₄
MW: 323.9200 g/mol
Bod tání: 318 °C
Storage Temperature: Ambient
MDL Number: MFCD00204593
CAS číslo: 13731-82-3
UN: 1759
ADR: 8 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR