Tisk…

N-(2,4-Dimethoxyphenyl)-2-chloroacetamide

Dodavatel: Apollo Scientific
Varování

Synonyms: N-(Chloroacetyl)-2,4-dimethoxyaniline

OR21448-1G OR21448-5G
APOSOR21448-1GEA 1660 CZK
APOSOR21448-1G APOSOR21448-5G
N-(2,4-Dimethoxyphenyl)-2-chloroacetamide
N-(2,4-Dimethoxyphenyl)-2-chloroacetamide
Vzorec: C₁₀H₁₂ClNO₃
Storage Temperature: Ambient
CAS číslo: 101908-41-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR