Tisk…

2-(Methylthio)nicotinamide

Dodavatel: Apollo Scientific

OR21420-1G
APOSOR21420-1GEA 2900 CZK
APOSOR21420-1G
2-(Methylthio)nicotinamide
2-(Methylthio)nicotinamide
Vzorec: C₇H₈N₂OS
Storage Temperature: Ambient
CAS číslo: 175135-28-1

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR