Tisk…

4-(Butylamino)benzoic acid 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: Tetracaine Related Compound B

OR19582-5G
APOSOR19582-5GEA 3440 CZK
APOSOR19582-5G
4-(Butylamino)benzoic acid 97%
4-(Butylamino)benzoic acid
Vzorec: C₁₁H₁₅NO₂
MW: 193.242 g/mol
Bod tání: 149…151 °C
Storage Temperature: Ambient
MDL Number: MFCD00002536
CAS číslo: 4740-24-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR