Tisk…

6-Bromopicolinic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: 6-Bromo-2-pyridinecarboxylic acid, 6-Bromopyridine-2-carboxylic acid

OR1906-25G OR1906-5G OR1906-100G
APOSOR1906-25GEA 1040 CZK
APOSOR1906-25G APOSOR1906-5G APOSOR1906-100G
6-Bromopicolinic acid
6-Bromopicolinic acid
Vzorec: C₆H₄BrNO₂
MW: 202.01
Bod tání: 188…194 °C
Storage Temperature: Ambient
MDL Number: MFCD00093197
CAS číslo: 21190-87-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR