Tisk…

5-(Pyridin-3-yl)-1,2,4-oxadiazol-3-amine

Dodavatel: Apollo Scientific

OR18583-1G OR18583-250MG OR18583-500MG
APOSOR18583-1GEA 6910 CZK
APOSOR18583-1G APOSOR18583-250MG APOSOR18583-500MG
5-(Pyridin-3-yl)-1,2,4-oxadiazol-3-amine
5-(Pyridin-3-yl)-1,2,4-oxadiazol-3-amine
Vzorec: C₇H₆N₄O
Storage Temperature: Ambient
MDL Number: MFCD09864898
CAS číslo: 73631-18-2

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR