Tisk…

(Isopropylthio)benzene 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: Isopropyl phenyl sulphide

OR18312-1KG OR18312-250G OR18312-50G
APOSOR18312-1KGEA 3480 CZK
APOSOR18312-1KG APOSOR18312-250G APOSOR18312-50G
(Isopropylthio)benzene 97%
(Isopropylthio)benzene
Vzorec: C₉H₁₂S
MW: 152.26
Bod varu: 105 °C/ 40mmHg
Bod tání: 56
Hustota: 0.9859
Storage Temperature: Refrigerator
MDL Number: MFCD00039842
CAS číslo: 3019-20-3
EINECS: 221-162-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR