Tisk…

Monomethyl-trans-1,4-cyclohexanedicarboxylate

Dodavatel: Apollo Scientific
Varování

Synonyms: Methyl Hydrogen trans-1,4-Cyclohexanedicarboxylate, trans-4-(Methoxycarbonyl)cyclohexanecarboxylic acid, trans-1,4-Cyclohexanedicarboxylic Acid Monomethyl Ester

OR17609-100G
APOSOR17609-100GEA 2400 CZK
APOSOR17609-100G
Monomethyl-trans-1,4-cyclohexanedicarboxylate
Monomethyl-trans-1,4-cyclohexanedicarboxylate
Vzorec: C₉H₁₄O₄
MW: 186.21 g/mol
Bod tání: 128…131 °C
Storage Temperature: Ambient
MDL Number: MFCD01311247
CAS číslo: 15177-67-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR