Tisk…

4-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid (cis and trans mixture) 98%

Dodavatel: Apollo Scientific

OR17545-5G OR17545-25G
APOSOR17545-5GEA 1210 CZK
APOSOR17545-5G APOSOR17545-25G
4-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid (cis and trans mixture) 98%
4-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid (cis and trans mixture)
Vzorec: C₁₂H₂₁NO₄
Storage Temperature: Ambient
MDL Number: MFCD03453262
CAS číslo: 130309-46-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR