Tisk…

3,7-Dihydro-5-iodo-4H-pyrrolo[2,3-d]pyrimidin-4-one

Dodavatel: Apollo Scientific
Varování

OR17053-250MG
APOSOR17053-250MGEA 622 CZK
APOSOR17053-250MG
3,7-Dihydro-5-iodo-4H-pyrrolo[2,3-d]pyrimidin-4-one
3,7-Dihydro-5-iodo-4H-pyrrolo[2,3-d]pyrimidin-4-one
Vzorec: C₆H₄IN₃O
Storage Temperature: Refrigerator
MDL Number: MFCD09746273
CAS číslo: 135352-71-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR