Tisk…

1'-Boc-1,2-dihydro-2-oxospiro[3H-indole-3,4'-piperidine]

Dodavatel: Apollo Scientific
Varování

OR16702-250MG
APOSOR16702-250MGEA 1620 CZK
APOSOR16702-250MG
1'-Boc-1,2-dihydro-2-oxospiro[3H-indole-3,4'-piperidine]
1'-Boc-1,2-dihydro-2-oxospiro[3H-indole-3,4'-piperidine]
Vzorec: C₁₇H₂₂N₂O₃
Storage Temperature: Ambient
MDL Number: MFCD10566051
CAS číslo: 252882-60-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR