Tisk…

4-[(Methylamino)methyl]-4-phenyltetrahydro-2H-pyran

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: N-methyl-1-(4-phenyltetrahydro-2H-pyran-4-yl)methylamine

OR16552-1G OR16552-250MG
APOSOR16552-1GEA 6220 CZK
APOSOR16552-1G APOSOR16552-250MG
4-[(Methylamino)methyl]-4-phenyltetrahydro-2H-pyran
4-[(Methylamino)methyl]-4-phenyltetrahydro-2H-pyran
Vzorec: C₁₃H₁₉NO
MW: 205.296 g/mol
Bod tání: 38…39.5 °C
Storage Temperature: Ambient
MDL Number: MFCD11841073
CAS číslo: 958443-30-6
UN: 3259
ADR: 8 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR