Tisk…

N-Methyl-1-(p-tolyl)methanamine

Dodavatel: Apollo Scientific
Nebezpečí

OR16470-1G OR16470-25G OR16470-5G
APOSOR16470-1GEA 1660 CZK
APOSOR16470-1G APOSOR16470-25G APOSOR16470-5G
N-Methyl-1-(p-tolyl)methanamine
N-Methyl-1-(p-tolyl)methanamine
Vzorec: C₉H₁₃N
Storage Temperature: Ambient
MDL Number: MFCD04115412
CAS číslo: 699-04-7
UN: 3265
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR