Tisk…

(S)-1-Methyl-3-(hydroxymethyl)piperidine

Dodavatel: Apollo Scientific
Varování

OR15996-1G OR15996-250MG OR15996-5G
APOSOR15996-1GEA 5560 CZK
APOSOR15996-1G APOSOR15996-250MG APOSOR15996-5G
(S)-1-Methyl-3-(hydroxymethyl)piperidine
(S)-1-Methyl-3-(hydroxymethyl)piperidine
Vzorec: C₇H₁₅NO
Storage Temperature: Ambient
MDL Number: MFCD11035736
CAS číslo: 205194-35-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR