Tisk…

3-Amino-7-azaindole dihydrochloride 95+%

Dodavatel: Apollo Scientific
Varování

Synonyms: 3-Amino-1H-pyrrolo[2,3-b]pyridine dihydrochloride, 1H-Pyrrolo[2,3-b]pyridin-3-amine dihydrochloride

OR15982-250MG
APOSOR15982-250MGEA 6720 CZK
APOSOR15982-250MG
3-Amino-7-azaindole dihydrochloride 95+%
3-Amino-7-azaindole dihydrochloride
Vzorec: C₇H₇N₃·2ClH
MW: 206,072 g/mol
Bod tání: 253 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD11035725
CAS číslo: 100960-08-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR