Tisk…

1-(4-Bromo-2-methylphenyl)-1H-pyrrole

Dodavatel: Apollo Scientific

OR15387-1G
APOSOR15387-1GEA 5130 CZK
APOSOR15387-1G
1-(4-Bromo-2-methylphenyl)-1H-pyrrole
1-(4-Bromo-2-methylphenyl)-1H-pyrrole
Vzorec: C₁₁H₁₀BrN
Storage Temperature: Ambient
MDL Number: MFCD02665256
CAS číslo: 383137-70-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR