Tisk…

2-Aminothiazole-4-carboxylic acid ethyl ester hydrobromide

Dodavatel: Apollo Scientific
Varování

OR14763-10G
APOSOR14763-10GEA 665 CZK
APOSOR14763-10G
2-Aminothiazole-4-carboxylic acid ethyl ester hydrobromide
2-Aminothiazole-4-carboxylic acid ethyl ester hydrobromide
Vzorec: C₆H₉BrN₂O₂S
Bod tání: 175…177 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06653384
CAS číslo: 127942-30-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR