Tisk…

2,4-Dichloro-D-phenylalanine

Dodavatel: Apollo Scientific
Varování

Synonyms: H-D-Phe(2,4-DiCl)-OH

OR14718-100MG
APOSOR14718-100MGEA 622 CZK
APOSOR14718-100MG
2,4-Dichloro-D-phenylalanine
2,4-Dichloro-D-phenylalanine
Vzorec: C₉H₉Cl₂NO₂
MW: 234.09 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01860873
CAS číslo: 114872-98-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR