Tisk…

3-Nitro-D-phenylalanine 97%

Dodavatel: Apollo Scientific

OR14708-1G
APOSOR14708-1GEA 1040 CZK
APOSOR14708-1G
3-Nitro-D-phenylalanine 97%
3-Nitro-D-phenylalanine
Vzorec: C₉H₁₀N₂O₄
Storage Temperature: Ambient
MDL Number: MFCD04117829
CAS číslo: 169530-97-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR