Tisk…

Fmoc-Thi-OH, Fmoc-3-L-Ala(3-Thienyl)-OH 97%

Dodavatel: Apollo Scientific

OR14648-1G OR14648-250MG OR14648-5G
APOSOR14648-1GEA 6750 CZK
APOSOR14648-1G APOSOR14648-250MG APOSOR14648-5G
Fmoc-Thi-OH, Fmoc-3-L-Ala(3-Thienyl)-OH 97%
Fmoc-Thi-OH, Fmoc-3-L-Ala(3-Thienyl)-OH
Vzorec: C₂₂H₁₉NO₄S
Storage Temperature: Refrigerator
MDL Number: MFCD00151916
CAS číslo: 186320-06-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR