Tisk…

2-(Chloromethyl)-1-(4-chlorophenoxy)-4-nitrobenzene

Dodavatel: Apollo Scientific

OR14097-10G OR14097-1G OR14097-5G
APOSOR14097-10GEA 47940 CZK
APOSOR14097-10G APOSOR14097-1G APOSOR14097-5G
2-(Chloromethyl)-1-(4-chlorophenoxy)-4-nitrobenzene
2-(Chloromethyl)-1-(4-chlorophenoxy)-4-nitrobenzene
Vzorec: C₁₃H₉Cl₂NO₃
Bod tání: 90…92 °C
Storage Temperature: Ambient
MDL Number: MFCD08689723
CAS číslo: 924862-19-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR