Tisk…

3-(2-Methoxycarbonylethyl)benzeneboronic acid 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: (3-(3-Methoxy-3-oxopropyl)phenyl)boronic acid

OR13161-1G OR13161-5G
APOSOR13161-1GEA 4600 CZK
APOSOR13161-1G APOSOR13161-5G
3-(2-Methoxycarbonylethyl)benzeneboronic acid 98%
3-(2-Methoxycarbonylethyl)benzeneboronic acid
Vzorec: C₁₀H₁₃BO₄
MW: 208.02 g/mol
Bod tání: 96…100 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04115648
CAS číslo: 833472-82-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR