Tisk…

3-(tert-Butylcarbamoyl)phenylboronic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: 3-(tert-Butylaminocarbonyl)phenylboronic acid

OR13122-1G OR13122-5G
APOSOR13122-1GEA 3310 CZK
APOSOR13122-1G APOSOR13122-5G
3-(tert-Butylcarbamoyl)phenylboronic acid
3-(tert-Butylcarbamoyl)phenylboronic acid
Vzorec: C₁₁H₁₆BNO₃
Bod tání: 120…126 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04115693
CAS číslo: 183158-30-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR