Tisk…

4-(2-Methoxyphenoxy)benzoic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-(4-Carboxyphenoxy)anisole

OR12951-250MG
APOSOR12951-250MGEA 1040 CZK
APOSOR12951-250MG
4-(2-Methoxyphenoxy)benzoic acid
4-(2-Methoxyphenoxy)benzoic acid
Vzorec: C₁₄H₁₂O₄
MW: 244.243 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01570262
CAS číslo: 103203-54-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR