Tisk…

4-(4-Methoxybenzyl)piperidine hydrochloride 95%

Dodavatel: Apollo Scientific
Varování

OR12871-1G OR12871-250MG OR12871-5G
APOSOR12871-1GEA 5630 CZK
APOSOR12871-1G APOSOR12871-250MG APOSOR12871-5G
4-(4-Methoxybenzyl)piperidine hydrochloride 95%
4-(4-Methoxybenzyl)piperidine hydrochloride
Vzorec: C₁₃H₁₉NO·ClH
Storage Temperature: Ambient
MDL Number: MFCD03840137
CAS číslo: 37581-27-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR