Tisk…

2-Phenyl-2-(pyrrolidin-1-yl)ethylamine dihydrochloride 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: 1-(2-Amino-1-phenylethyl)pyrrolidine dihydrochloride, β-(Pyrrolidin-1-yl)phenethylamine dihydrochloride

OR12694-250MG OR12694-1G
APOSOR12694-250MGEA 14490 CZK
APOSOR12694-250MG APOSOR12694-1G
2-Phenyl-2-(pyrrolidin-1-yl)ethylamine dihydrochloride 95%
2-Phenyl-2-(pyrrolidin-1-yl)ethylamine dihydrochloride
Vzorec: C₁₂H₁₈N₂·2ClH
MW: 263.207 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08445576
CAS číslo: 31788-96-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR