Tisk…

5-Methylthiophene-2-carbonyl chloride

Dodavatel: Apollo Scientific
Nebezpečí

OR12300-1G OR12300-250MG OR12300-100MG OR12300-500MG OR12300-2.5G OR12300-5G
APOSOR12300-1GEA 7440 CZK
APOSOR12300-1G APOSOR12300-250MG APOSOR12300-100MG APOSOR12300-500MG APOSOR12300-2.5G APOSOR12300-5G
5-Methylthiophene-2-carbonyl chloride
5-Methylthiophene-2-carbonyl chloride
Vzorec: C₆H₅ClOS
Bod varu: 120 °C/8mmHg
Storage Temperature: Ambient
MDL Number: MFCD00130092
CAS číslo: 31555-59-6
UN: 3265
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR