Tisk…

6-Methyl-2H-1,4-benzoxazine-2,3(4H)-dione 3-oxime

Dodavatel: Apollo Scientific

OR12122-10G OR12122-1G OR12122-5G
APOSOR12122-10GEA 25460 CZK
APOSOR12122-10G APOSOR12122-1G APOSOR12122-5G
6-Methyl-2H-1,4-benzoxazine-2,3(4H)-dione 3-oxime
6-Methyl-2H-1,4-benzoxazine-2,3(4H)-dione 3-oxime
Vzorec: C₉H₈N₂O₃
Storage Temperature: Ambient
MDL Number: MFCD08689791
CAS číslo: 937602-17-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR