Tisk…

1-[1-(tert-Butoxycarbonyl)-4-piperidinyl]-5-oxo-3-pyrrolidinecarboxylic acid

Dodavatel: Apollo Scientific
Varování

OR12054-1G
APOSOR12054-1GEA 8070 CZK
APOSOR12054-1G
1-[1-(tert-Butoxycarbonyl)-4-piperidinyl]-5-oxo-3-pyrrolidinecarboxylic acid
1-[1-(tert-Butoxycarbonyl)-4-piperidinyl]-5-oxo-3-pyrrolidinecarboxylic acid
Vzorec: C₁₅H₂₄N₂O₅
MW: 312.37 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04973994
CAS číslo: 937601-51-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR