Tisk…

8-Bromo-6-nitroimidazo[1,2-a]pyridine hydrochloride

Dodavatel: Apollo Scientific
Varování

OR11664-1G
APOSOR11664-1GEA 953 CZK
APOSOR11664-1G
8-Bromo-6-nitroimidazo[1,2-a]pyridine hydrochloride
8-Bromo-6-nitroimidazo[1,2-a]pyridine hydrochloride
Vzorec: C₇H₅BrClN₃O₂
Storage Temperature: Ambient
MDL Number: MFCD09800973
CAS číslo: 957120-43-3

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR