Tisk…

2-Amino-4-(ethoxycarbonyl)benzeneboronic acid hydrochloride 95%

Dodavatel: Apollo Scientific

OR11454-1G
APOSOR11454-1GEA 18920 CZK
APOSOR11454-1G
2-Amino-4-(ethoxycarbonyl)benzeneboronic acid hydrochloride 95%
2-Amino-4-(ethoxycarbonyl)benzeneboronic acid hydrochloride
Vzorec: C₉H₁₂BNO₄·ClH
MDL Number: MFCD09475809
CAS číslo: 957034-59-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR