Tisk…

Ethyl (4-amino-3,5-dibromophenyl)acetate

Dodavatel: Apollo Scientific
Varování

OR111424-1G
APOSOR111424-1GEA 8200 CZK
APOSOR111424-1G
Ethyl (4-amino-3,5-dibromophenyl)acetate
Ethyl (4-amino-3,5-dibromophenyl)acetate
Vzorec: C₁₀H₁₁Br₂NO₂
MW: 337.01 g/mol
MDL Number: MFCD12547823
CAS číslo: 122063-99-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR