Tisk…

4-Methoxy-2,3,6-trimethylbenzenesulphonyl chloride 95%

Dodavatel: Apollo Scientific

OR111378-1G
APOSOR111378-1GEA 2530 CZK
APOSOR111378-1G
4-Methoxy-2,3,6-trimethylbenzenesulphonyl chloride 95%
4-Methoxy-2,3,6-trimethylbenzenesulphonyl chloride
Vzorec: C₁₀H₁₃ClO₃S
MW: 248,73 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00038846
CAS číslo: 80745-07-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR