Tisk…

Pyrido[3',2':4,5]thieno[3,2-d]pyrimidin-4(3H)-one

Dodavatel: Apollo Scientific

OR111011-1G
APOSOR111011-1GEA 8270 CZK
APOSOR111011-1G
Pyrido[3',2':4,5]thieno[3,2-d]pyrimidin-4(3H)-one
Pyrido[3',2':4,5]thieno[3,2-d]pyrimidin-4(3H)-one
Vzorec: C₉H₅N₃OS
Storage Temperature: Ambient
MDL Number: MFCD01312791
CAS číslo: 55115-79-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR