Tisk…

1-Bromo-4-(isopropylthio)benzene

Dodavatel: Apollo Scientific
Varování

OR111003-1G OR111003-100MG OR111003-5G
APOSOR111003-1GEA 3740 CZK
APOSOR111003-1G APOSOR111003-100MG APOSOR111003-5G
1-Bromo-4-(isopropylthio)benzene
1-Bromo-4-(isopropylthio)benzene
Vzorec: C₉H₁₁BrS
Storage Temperature: Ambient
MDL Number: MFCD20484423
CAS číslo: 70398-89-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR