Tisk…

5-Amino-2-diisopropylamino-1,3,4-thiadiazole

Dodavatel: Apollo Scientific

Synonyms: N,N-Diisopropyl-1,3,4-thiadiazole-2,5-diamine

OR110699-1G OR110699-5G
APOSOR110699-1GEA 5290 CZK
APOSOR110699-1G APOSOR110699-5G
5-Amino-2-diisopropylamino-1,3,4-thiadiazole
5-Amino-2-diisopropylamino-1,3,4-thiadiazole
Vzorec: C₈H₁₆N₄S
MW: 200.31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00036052
CAS číslo: 72269-92-2

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR